Learn More
Medchemexpress LLC Ibuprofen-d4 | 15687-27-1 | 99.3% | C13H14D4O2 | 5 MG
SDP
Catalog No. 5000434860
Encompass_Preferred
Ibuprofen-d4 is a deuterium-labeled Ibuprofen, a potent, orally active, selective COX-1 inhibitor with an IC50 value of 13 μM. It inhibits cell proliferation, angiogenesis, and induces cell apoptosis, functioning as a nonsteroidal anti-inflammatory agent and a nitric oxide (NO) donor. It can be used in the research of pain, swelling, inflammation, infection, immunology, and cancers.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling has the potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-34860 Supplier Medchemexpress LLC Supplier No. HY78131S25MG
Ibuprofen-d4 is a deuterium-labeled Ibuprofen, a potent, orally active, selective COX-1 inhibitor with an IC50 value of 13 μM. It inhibits cell proliferation, angiogenesis, and induces cell apoptosis, functioning as a nonsteroidal anti-inflammatory agent and a nitric oxide (NO) donor. It can be used in the research of pain, swelling, inflammation, infection, immunology, and cancers.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling has the potential to affect the pharmacokinetic and metabolic profiles of drugs