Learn More
Ibuprofen Impurity I, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111003681 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
Catalog No. Quantity
11-100-3681 50 mg
1 options
Catalog No. 11-100-3681 Supplier MilliporeSigma Supelco Supplier No. PHR198450MG
Add to Cart
Add to Cart
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 mg
Grade Certified Reference Material