Learn More
AdipoGen Ibuprofen Impurity K
SDP
Catalog No. 502560626
Encompass_Preferred
Chemical. CAS 43153-07-7. Formula C10H10O3. MW 178.18. The metabolite 2RS-2-4-Formylphenylpropionic acid Ibuprofen Impurity K is a degradation product arising from oxidative and thermal treatments of Ibuprofen which is a non-steroidal anti-inflammatory drug NSAID used to relieve pain from various conditions such as headache, dental pain or menstrual cramps. Ibuprofen is an anti-inflammatory inhibitor targeting COX-1 and COX-2. This product can be used as a working standard or secondary analytical reference standard. It can also be used as an organic building block and intermediate for synthesis.
Catalog No. 50-256-0626 Supplier AdipoGen Supplier No. CDXI0303M100
Chemical. CAS 43153-07-7. Formula C10H10O3. MW 178.18. The metabolite 2RS-2-4-Formylphenylpropionic acid Ibuprofen Impurity K is a degradation product arising from oxidative and thermal treatments of Ibuprofen which is a non-steroidal anti-inflammatory drug NSAID used to relieve pain from various conditions such as headache, dental pain or menstrual cramps. Ibuprofen is an anti-inflammatory inhibitor targeting COX-1 and COX-2. This product can be used as a working standard or secondary analytical reference standard. It can also be used as an organic building block and intermediate for synthesis.