Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ibuprofen Impurity O Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015044 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5044 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5044 Supplier MilliporeSigma Supelco Supplier No. PHR198330MG
Add to Cart
Add to Cart

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

TRUSTED_SUSTAINABILITY

Specifications

CAS 64451-76-9
Linear Formula C13H18O2
Synonym 2-[4-(1-Methylpropyl)-phenyl]propanoic acid; 2-[4-(1-methylpropyl)phenyl]propanoic acid
Type Certified Reference Material
Quantity 30 mg
Formula Weight 206.28
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.