Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ibuprofen Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013258 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3258 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3258 Supplier MilliporeSigma Supelco Supplier No. PHR10041G
Add to Cart
Add to Cart
Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 15687-27-1
Physical Form Neat
Linear Formula C13H18O2
MDL Number MFCD00010393
Synonym alpha-Methyl-4-(isobutyl)phenylacetic acid; (+/-)-2-(4-Isobutylphenyl)propanoic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 206.28
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.