Learn More
American Radiolabeled Chemicals Inc IBUPROFEN RS-3H(G) 250UC
SDP
Catalog No. 5090108058
Encompass_Preferred
Ibuprofen, RS-3H(G) 250uCi in ethanol, 1 mCi/ml, S.A. 1-5 Ci/mmol, M.W. 206.27, Exclusive
Catalog No. 50-901-08058 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 0392250UC
Ibuprofen, RS-3H(G) 250uCi in ethanol, 1 mCi/ml, S.A. 1-5 Ci/mmol, M.W. 206.27, Exclusive