Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ibuprofen sodium salt, 98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013894 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3894 100 g
11-101-3893 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3894 Supplier MilliporeSigma Supelco Supplier No. I1892100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ibuprofen belongs to the class of non-steroidal anti-inflammatory drugs, widely used for the treatment of mild to moderate pain and several forms of arthritis.

Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 31121-93-4
Physical Form Neat
Linear Formula C13H17O2 Na
MDL Number MFCD00133414
Synonym alpha-Methyl-4-(isobutyl)phenylacetic acid
Solubility Soluble in water
Type Analytical Standard
Quantity 100 g
Formula Weight 228.26
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.