Learn More
Medchemexpress LLC Ibuprofen standard | 15687-27-1 | 99.95% | C13H18O2 | 50 MG
SDP
Catalog No. 5000434859
Encompass_Preferred
Ibuprofen Standard is an analytical standard intended for research and analytical applications. It is a potent, orally active, selective COX-1 inhibitor that inhibits cell proliferation, angiogenesis, and induces cell apoptosis. This nonsteroidal anti-inflammatory agent and nitric oxide (NO) donor is commonly used in qualitative, quantitative, and methodological research experiments.
  • Potent, orally active, selective COX-1 inhibitor
  • Inhibits cell proliferation, angiogenesis, and induces cell apoptosis
  • Nonsteroidal anti-inflammatory agent and nitric oxide (NO) donor
  • Used in research related to pain, swelling, inflammation, infection, immunology, and cancers
  • Commonly used in HPLC, GC, and MS techniques
Catalog No. 50-004-34859 Supplier Medchemexpress LLC Supplier No. HY78131R50MG
Ibuprofen Standard is an analytical standard intended for research and analytical applications. It is a potent, orally active, selective COX-1 inhibitor that inhibits cell proliferation, angiogenesis, and induces cell apoptosis. This nonsteroidal anti-inflammatory agent and nitric oxide (NO) donor is commonly used in qualitative, quantitative, and methodological research experiments.
  • Potent, orally active, selective COX-1 inhibitor
  • Inhibits cell proliferation, angiogenesis, and induces cell apoptosis
  • Nonsteroidal anti-inflammatory agent and nitric oxide (NO) donor
  • Used in research related to pain, swelling, inflammation, infection, immunology, and cancers
  • Commonly used in HPLC, GC, and MS techniques