Learn More
Apexbio Technology LLC Ibutamoren Mesylate 159752-10-0 50mg
SDP
Catalog No. 5000093630
Encompass_Preferred
Ibutamoren mesylate (CAS 159752-10-0) also known as MK-0677 is an orally bioavailable nonpeptide growth hormone (GH) secretagogue It acts by stimulating GH release via activation of ghrelin receptors located in the pituitary and hypothalamus distinct from the classical GH-releasing hormone receptor Administration of ibutamoren mesylate results in elevated plasma concentrations of growth hormone and insulin-like growth factor 1 (IGF-1) without significantly affecting cortisol levels This compound is under investigation for its ability to enhance muscle mass and bone mineral density and is being studied as a potential therapy for conditions involving GH deficiency or age-related frailty
Catalog No. 50-000-93630 Supplier Apexbio Technology LLC Supplier No. A348150
Ibutamoren mesylate (CAS 159752-10-0) also known as MK-0677 is an orally bioavailable nonpeptide growth hormone (GH) secretagogue It acts by stimulating GH release via activation of ghrelin receptors located in the pituitary and hypothalamus distinct from the classical GH-releasing hormone receptor Administration of ibutamoren mesylate results in elevated plasma concentrations of growth hormone and insulin-like growth factor 1 (IGF-1) without significantly affecting cortisol levels This compound is under investigation for its ability to enhance muscle mass and bone mineral density and is being studied as a potential therapy for conditions involving GH deficiency or age-related frailty