Learn More
Medchemexpress LLC Ibutamoren Mesylate | 159752-10-0 | 99.83% | C28H40N4O8S2 | 50 MG
Supplier: Medchemexpress LLC HY5084450MG
Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. It is an orally active growth hormone (GH) secretagogue.
- Potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist.
- Orally active growth hormone (GH) secretagogue.
- Increases body weight gain and increases serum IGF-1 and GH levels in dogs (at 5 mg/kg/day).
- Chronic oral administration increases and maintains elevated IGF-I levels.
- Significantly increases peak GH concentrations after oral administration.
- Induces an immediate, large, long-lasting increase in GH levels when administered orally or i.v.