Learn More
Medchemexpress LLC Ibutamoren Mesylate | 159752-10-0 | 99.86% | C28H40N4O8S2 | 1 G
SDP
Catalog No. 5000357991
Encompass_Preferred
Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. It is an orally active growth hormone (GH) secretagogue.
  • Potent, non-peptide growth hormone secretagogue receptor (GHSR) agonist
  • Orally active growth hormone (GH) secretagogue
  • For research use only
Catalog No. 50-003-57991 Supplier Medchemexpress LLC Supplier No. HY508441G
Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. It is an orally active growth hormone (GH) secretagogue.
  • Potent, non-peptide growth hormone secretagogue receptor (GHSR) agonist
  • Orally active growth hormone (GH) secretagogue
  • For research use only