Learn More
Medchemexpress LLC Ibutamoren (Mesylate) | 159752-10-0 | C28H40N4O8S2 | 100 MG
SDP
Catalog No. 5000359153
Encompass_Preferred
Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. It is an orally active growth hormone (GH) secretagogue, shown to increase serum IGF-1 and GH levels in vivo.
  • Potent, non-peptide growth hormone secretagogue receptor (GHSR) agonist
  • Orally active growth hormone (GH) secretagogue
  • Increases serum IGF-1 and GH levels
  • Increases peak GH concentrations after oral administration
Catalog No. 50-003-59153 Supplier Medchemexpress LLC Supplier No. HY50844100MG
Ibutamoren Mesylate (MK-677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. It is an orally active growth hormone (GH) secretagogue, shown to increase serum IGF-1 and GH levels in vivo.
  • Potent, non-peptide growth hormone secretagogue receptor (GHSR) agonist
  • Orally active growth hormone (GH) secretagogue
  • Increases serum IGF-1 and GH levels
  • Increases peak GH concentrations after oral administration