Learn More
TARGETMOL CHEMICALS INC Ibutamoren Mesylate 50MG
SDP
Catalog No. 502877898
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Ibutamoren Mesylate (MK-0677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. Purity 97.98%
Catalog No. 50-287-7898 Supplier TARGETMOL CHEMICALS INC Supplier No. T711250MG
Also available in 1 mL, 5 mg, 10 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Ibutamoren Mesylate (MK-0677) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. Purity 97.98%