Learn More
Medchemexpress LLC HY-50844 500mg , Ibutamoren (Mesylate) MK-677;MK-0677 CAS:159752-10-0 Purity:98%
SDP
Catalog No. 501492450
Encompass_Preferred
Medchemexpress, HY-50844 500mg Ibutamoren (Mesylate) MK-677;MK-0677 CAS:159752-10-0 Formula:C28H40N4O8S2 Purity:98% Ibutamoren (Mesylate) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2450 Supplier Medchemexpress LLC Supplier No. HY50844500MG
Medchemexpress, HY-50844 500mg Ibutamoren (Mesylate) MK-677;MK-0677 CAS:159752-10-0 Formula:C28H40N4O8S2 Purity:98% Ibutamoren (Mesylate) is a potent, non-peptide Growth hormone secretagogue receptor (GHSR) agonist. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.