Learn More
Cayman Chemical Ibutamorenmesylate 50mg
SDP
Catalog No. 502966941
Encompass_Preferred
An orally-active, non-peptidic agonist of GHSR (Kd = 0.4 nM) and, as a result, is a GH secretagogue; elevates GH in dogs after oral doses as low as 0.125 mg/kg, without significantly changing plasma levels of aldosterone, luteinizing hormone, thyroxine, or prolactin
Catalog No. 50-296-6941 Supplier Cayman Chemical Supplier No. 1800350 MG
An orally-active, non-peptidic agonist of GHSR (Kd = 0.4 nM) and, as a result, is a GH secretagogue; elevates GH in dogs after oral doses as low as 0.125 mg/kg, without significantly changing plasma levels of aldosterone, luteinizing hormone, thyroxine, or prolactin