Learn More
Apexbio Technology LLC Ibutilide Fumarate 122647-32-9 10mg
SDP
Supplier:  Apexbio Technology LLC B227810
Encompass_Preferred
Ibutilide Fumarate (CAS 122647-32-9) is a small molecule classified as a Class III antiarrhythmic agent It acts primarily by enhancing slow inward sodium currents and delaying repolarization through inhibition of the rapid component of the cardiac delayed rectifier potassium current (IKr) resulting in prolongation of the cardiac action potential Ibutilide is utilized in research to investigate pharmacological cardioversion of atrial arrhythmias specifically studying the acute conversion of atrial fibrillation and atrial flutter to normal sinus rhythm Its properties make it valuable for exploring the mechanisms of antiarrhythmic therapy and cardiac electrophysiology
Catalog No. 50-224-0635