Learn More
Medchemexpress LLC Ibutilide (fumarate) | 122647-32-9 | 99.7% | 100 MG
SDP
Catalog No. 5000406109
Encompass_Preferred
Ibutilide fumarate is an action potential-prolonging antiarrhythmic agent. It is a potent blocker of the rapidly activating delayed rectifier K+ current (IKr) in AT-1 cells, playing a role in cardiac repolarization. This product is suitable for research applications in cardiac physiology and pharmacology.
  • Potent IKr blocker
  • Prolongs cardiac repolarization
  • Relevant for studies on atrial tumor myocytes
  • Useful in research related to congestive heart failure models
Catalog No. 50-004-06109 Supplier Medchemexpress LLC Supplier No. HYB0387100MG
Ibutilide fumarate is an action potential-prolonging antiarrhythmic agent. It is a potent blocker of the rapidly activating delayed rectifier K+ current (IKr) in AT-1 cells, playing a role in cardiac repolarization. This product is suitable for research applications in cardiac physiology and pharmacology.
  • Potent IKr blocker
  • Prolongs cardiac repolarization
  • Relevant for studies on atrial tumor myocytes
  • Useful in research related to congestive heart failure models