Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ibutilide (fumarate) | 122647-32-9 | 99.7% | 5 MG
SDP

Catalog No. 5000406424
Encompass_Preferred

Ibutilide fumarate (U70226E) is an action potential-prolonging antiarrhythmic agent. It acts as a potent blocker of the rapidly activating delayed rectifier K⁺ current (IKr) in AT-1 cells. This compound is intended for research use only.

  • Potent IKr blocker with an EC50 value of 20 nM in AT-1 cells.
  • Blocks IKr in cells expressing HERG+MDR1*1 to the same extent as HERG alone.
  • Prolongs cardiac repolarization in vitro and in vivo.
  • Appears as a white to off-white solid.
  • Recommended storage for solid: 4°C, sealed, away from moisture.
  • Recommended storage in solvent: -80°C for 6 months, or -20°C for 1 month.

Catalog No. 50-004-06424 Supplier Medchemexpress LLC Supplier No. HYB03875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ibutilide fumarate (U70226E) is an action potential-prolonging antiarrhythmic agent. It acts as a potent blocker of the rapidly activating delayed rectifier K⁺ current (IKr) in AT-1 cells. This compound is intended for research use only.

  • Potent IKr blocker with an EC50 value of 20 nM in AT-1 cells.
  • Blocks IKr in cells expressing HERG+MDR1*1 to the same extent as HERG alone.
  • Prolongs cardiac repolarization in vitro and in vivo.
  • Appears as a white to off-white solid.
  • Recommended storage for solid: 4°C, sealed, away from moisture.
  • Recommended storage in solvent: -80°C for 6 months, or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.