Learn More
Selleck Chemical LLC Ibutilide Fumarate S2118-10mg
SDP
Catalog No. 503109604
Encompass_Preferred
Ibutilide Fumarate (U-70226E Corvert Fumarate) is a Class III antiarrhythmic agent that is indicated for acute cardioconversion of atrial fibrillation and atrial flutter of a recent onset to sinus rhythm by induction of slow inward sodium current which prolongs action potential and refractory period of myocardial cells
Catalog No. 50-310-9604 Supplier Selleck Chemical LLC Supplier No. S211810MG
Ibutilide Fumarate (U-70226E Corvert Fumarate) is a Class III antiarrhythmic agent that is indicated for acute cardioconversion of atrial fibrillation and atrial flutter of a recent onset to sinus rhythm by induction of slow inward sodium current which prolongs action potential and refractory period of myocardial cells