Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ibutilide Hemifumarate 98.0+%, TCI America™
SDP

Catalog No. I094620MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I094620MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I094620MG Supplier TCI America Supplier No. I094620MG

Chemical Identifiers

CAS 122647-32-9
Molecular Formula C44H76N4O10S2
Molecular Weight (g/mol) 885.23
MDL Number MFCD01715410
InChI Key PCIOHQNIRPWFMV-WXXKFALUNA-N
Synonym ibutilide fumarate, corvert, ibutilide fumarate usan, ibutilide hemifumarate, ibutilide hemifumarate salt, corvert tn, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfoanilide e-2-butenedioate 2:1, +--4'-4-ethylheptylamino-1-hydroxybutyl methanesulfonanilide fumarate 2:1 salt, +--n-4-4-ethylheptylamino-1-hydroxybutyl phenyl methanesulfonamide e-butenedioate
PubChem CID 5281065
IUPAC Name (2E)-but-2-enedioic acid; bis(N-(4-{4-[ethyl(heptyl)amino]-1-hydroxybutyl}phenyl)methanesulfonamide)
SMILES OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1.CCCCCCCN(CC)CCCC(O)C1=CC=C(NS(C)(=O)=O)C=C1

Specifications

Color White
Quantity 20 mg
Formula Weight 442.62
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ibutilide Hemifumarate
RTECSNumber PB0475700
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.