Learn More
Cayman Chemical (±)-Ibutilide (hemifumarate), Purity: ≥98%, CAS: 122647-32-9, 50 mg
SDP
Catalog No. 502972415
Encompass_Preferred
A class III antiarrhythmic agent; inhibits IKr in AT-1 myocytes (IC50 = 20 nM); enhances INa and increases the action potential duration in isolated guinea pig ventricular cells; decreases pinacidil-induced ventricular fibrillation in Langendorff isolated perfused rabbit hearts from 3-30 µM; increases the ERP of the left and right atrium in anesthetized pigs at 15 µg/kg, i.v.; prevents rapid pacing-induced atrial flutter in dogs from 0.25-5 mg/kg
Catalog No. 50-297-2415 Supplier Cayman Chemical Supplier No. 1501150 MG
A class III antiarrhythmic agent; inhibits IKr in AT-1 myocytes (IC50 = 20 nM); enhances INa and increases the action potential duration in isolated guinea pig ventricular cells; decreases pinacidil-induced ventricular fibrillation in Langendorff isolated perfused rabbit hearts from 3-30 µM; increases the ERP of the left and right atrium in anesthetized pigs at 15 µg/kg, i.v.; prevents rapid pacing-induced atrial flutter in dogs from 0.25-5 mg/kg