Learn More
DIGI KEY CORPORATION IBUTTON Temperature Logger
SDP
Catalog No. NC0839180
Encompass
IBUTTON Temperature Logger, Operating Temperature -40°C ~ 85°C, Memory Size 512B, 8KB
Catalog No. NC0839180 Supplier DIGI KEY CORPORATION Supplier No. DS1922LF5#ND
IBUTTON Temperature Logger, Operating Temperature -40°C ~ 85°C, Memory Size 512B, 8KB