Learn More
Sigma Aldrich Fine Chemicals Biosciences IBX polystyrene Novabioche
Catalog No. 502908854
Encompass
This polymer-bound version of IBX cleanly converts primary and secondary alcohols to the corresponding aldehydes and ketones [1] 2. It is particularly useful for the synthesis of peptide aldehydes as it oxidises peptide alcohols efficiently and spent resin can be simply removed by filtration.The product number for this product was previously 01-64-0445.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.
Catalog No. 50-290-8854 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8550430025
Add to Cart
Add to Cart
This polymer-bound version of IBX cleanly converts primary and secondary alcohols to the corresponding aldehydes and ketones [1] 2. It is particularly useful for the synthesis of peptide aldehydes as it oxidises peptide alcohols efficiently and spent resin can be simply removed by filtration.The product number for this product was previously 01-64-0445.To obtain a certificate of analysis (CoA) of a lot that begins with the letter A please select the option in the right hand menu Request a COA for Lots starting with A.