Learn More
Inorganic Ventures 1000µg/mL CARBONATE 125mL
SDP
Catalog No. 50723567
Encompass_Preferred
Matrix: Water (H2O), Starting Material: Na2CO3, Analyte: CO3-2 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-567 Supplier Inorganic Ventures Supplier No. ICCO31125ML
Matrix: Water (H2O), Starting Material: Na2CO3, Analyte: CO3-2 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search