Learn More
Inorganic Ventures 1000µg/mL FLUORIDE 125mL
SDP
Catalog No. 50723579
Encompass_Preferred
Matrix: Water (H2O), Starting Material: NaF, Analyte: F- 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-579 Supplier Inorganic Ventures Supplier No. ICF1125ML
Matrix: Water (H2O), Starting Material: NaF, Analyte: F- 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search