Learn More
Inorganic Ventures 1000µg/mL NITRITE 500mL
SDP
Catalog No. 50723621
Encompass_Preferred
Matrix: Water (H2O), Starting Material: NaNO2, Analyte: NO2- 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-621 Supplier Inorganic Ventures Supplier No. ICNO21500ML
Matrix: Water (H2O), Starting Material: NaNO2, Analyte: NO2- 1000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search