Learn More
Inorganic Ventures 1000µg/mL TARTRATE 125mL
SDP
Catalog No. 50723657
Encompass_Preferred
Inorganic Ventures' 1000 ppm (µg/mL) Tartrate for IC is a 125mL certified reference material set in a water matrix for stability. This tartrate standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-657 Supplier Inorganic Ventures Supplier No. ICTRTR1125ML
Inorganic Ventures' 1000 ppm (µg/mL) Tartrate for IC is a 125mL certified reference material set in a water matrix for stability. This tartrate standard is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search