Learn More
Abcam IC 261, CK1delta and CK1epsilon inhibitor, 25MG
Catalog No. 502705417
Encompass_Preferred
MW 311.3 Da, Purity >99%. Selective CK1δ and CK1ε inhibitor. Potent microtubulin polymerization inhibitor. Induces cell cycle arrest and apoptosis. Selectively inhibits tumor cells growth. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5417 Supplier Abcam Supplier No. AB14518925MG
Add to Cart
Add to Cart
MW 311.3 Da, Purity >99%. Selective CK1δ and CK1ε inhibitor. Potent microtubulin polymerization inhibitor. Induces cell cycle arrest and apoptosis. Selectively inhibits tumor cells growth. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement