Learn More
Abcam IC 261, CK1delta and CK1epsilon inhibitor, 5MG
Catalog No. 502705416
Encompass_Preferred
MW 311.3 Da, Purity >99%. Selective CK1δ and CK1ε inhibitor. Potent microtubulin polymerization inhibitor. Induces cell cycle arrest and apoptosis. Selectively inhibits tumor cells growth. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5416 Supplier Abcam Supplier No. AB1451895MG
Add to Cart
Add to Cart
MW 311.3 Da, Purity >99%. Selective CK1δ and CK1ε inhibitor. Potent microtubulin polymerization inhibitor. Induces cell cycle arrest and apoptosis. Selectively inhibits tumor cells growth. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement