Learn More
Medchemexpress LLC HY-10110 5mg Medchemexpress, IC-87114 CAS:371242-69-2 Purity:>98%
SDP
Catalog No. 501932038
Encompass_Preferred
Medchemexpress, HY-10110 5mg IC-87114 CAS:371242-69-2 IC-87114 is a potent and selective PI3Kδ inhibitor with IC 50 of 0.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2038 Supplier Medchemexpress LLC Supplier No. HY101105MG
Medchemexpress, HY-10110 5mg IC-87114 CAS:371242-69-2 IC-87114 is a potent and selective PI3Kδ inhibitor with IC 50 of 0.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.