Learn More
Waters Corp Acrodisc Syringe Filter, PES, 13 mm, 0.2 micron
Supplier:  Waters Corp WAT200844
Encompass_Preferred
Polyethersulfone (PES) IC filters are certified for low ionic backgrounds, ideal for ion chromatography applications. These versatile syringe filters support scientific and industrial uses, including liquid chromatography and pre-filtration.
  • Certified to contain low ionic backgrounds for ion chromatography.
  • Tested using a highly sensitive IC protocol to monitor inorganic extractables.
  • Certified for low levels of inorganic extractables, typically <20ppb for chloride, 6ppb for nitrate, 1ppb for phosphate, and 10ppb for sulfate.
  • Wide range of applications including biological sciences, pharmaceutical procedures, research and development, quality control, and environmental analysis.
  • Supports high-pressure liquid chromatography (HPLC), ultra-high-pressure liquid chromatography (UHPLC), and pre-filtration.
  • Recommended for use with the ACQUITY UPLC System to maintain peak performance.
Catalog No. 50-786-174