Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IC Fixation Buffer

Catalog No. FB001
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB001 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FB001 Supplier Invitrogen™ Supplier No. FB001
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Intracellular fixation buffer for use in flow cytomerty, particularly for fixation in intracellular staining procedures.

Intracellular fixation buffer for use in flow cytomerty, particularly for fixation in intracellular staining procedures. Composition is paraformaldehyde in phosphate buffered saline, pH 7.3.

These Made-to-Order catalog items have a 2 week lead time.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 2°C to 8°C. Do not freeze.
Product Type Fixation Buffer
Quantity 100 mL
Solution Type Fixation Buffer
For Use With (Application) Cell Surface Antibodies, Intracellular Cytokine Antibodies
pH 7.3
Shipping Condition Wet Ice
What fixative should I use to fix my cells or tissues for antibody labeling?

Aldehyde-based fixatives (e.g., formaldehyde, glutaraldehyde) crosslink various cellular components, which helps retain proteins and cell morphology, but some antigens can be masked by the crosslinking, requiring antigen retrieval methods to unmask the antigen binding sites. Also, aldehyde-based fixation requires permeabilization to allow entry of antibodies. Organic solvents such as methanol and acetone condense proteins and permeabilize the cells, but cell morphology can be compromised. Surface antigens can be labeled without fixation.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.