Learn More
R&R Lotions Pregloving Cleanroom Lotion, 32 oz Pump Bottle, White
SDP
Catalog No. NC9188536
Encompass
This item must be purchased in multiples of 12 per supplier requirements. This non-petroleum, non-contaminating and greaseless formula absorbs immediately into the skin without interfering with grip or dexterity. Contains no contaminates such as silicones, lanolins, mineral oils and glycerins. Fragrance and dye free with Vitamins A, D and E, this natural formula helps to relieve dermatitis, urticaria, skin flaking, chapping, irritation and sensitivity cause by regular use of nitrile and latex gloves.
Catalog No. NC9188536 Supplier R&R Lotions Supplier No. ICL32CR
This item must be purchased in multiples of 12 per supplier requirements. This non-petroleum, non-contaminating and greaseless formula absorbs immediately into the skin without interfering with grip or dexterity. Contains no contaminates such as silicones, lanolins, mineral oils and glycerins. Fragrance and dye free with Vitamins A, D and E, this natural formula helps to relieve dermatitis, urticaria, skin flaking, chapping, irritation and sensitivity cause by regular use of nitrile and latex gloves.