Learn More
Waters Corp Waters IC-Pak Anion Concentrator Inserts, Anion-Exchange, Ion-Exchange, Spherical
Catalog No. 50784965 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters offers IC-Pak resin-based columns that enable the analysis of a wide range of ions from various sample matrices, including both simple and complex ones. These columns provide an exceptional linear loading range from less than 1.0 ppb to over 400 ppm without dilution or pH limitations on the eluent or sample. This flexibility ensures accurate and reproducible anion and cation analyses at all concentration levels.
  • Designed for anion-exchange chemistry.
  • Utilizes ion-exchange separation mode.
  • Features spherical particle shape.
  • Operates effectively in HPLC systems.
  • Maximum pressure of 1000 psi (70 Bar).
  • pH range of 1 to 12.
  • Normal operating temperature at 25°C, with limits from 10°C to 50°C.
  • Compatible with 12% acetonitrile in water.
  • Packaged as 5 units per package.
Catalog No. 50-784-965 Supplier Waters Corp Supplier No. WAT007358
Waters offers IC-Pak resin-based columns that enable the analysis of a wide range of ions from various sample matrices, including both simple and complex ones. These columns provide an exceptional linear loading range from less than 1.0 ppb to over 400 ppm without dilution or pH limitations on the eluent or sample. This flexibility ensures accurate and reproducible anion and cation analyses at all concentration levels.
  • Designed for anion-exchange chemistry.
  • Utilizes ion-exchange separation mode.
  • Features spherical particle shape.
  • Operates effectively in HPLC systems.
  • Maximum pressure of 1000 psi (70 Bar).
  • pH range of 1 to 12.
  • Normal operating temperature at 25°C, with limits from 10°C to 50°C.
  • Compatible with 12% acetonitrile in water.
  • Packaged as 5 units per package.