Learn More
Waters Corp Waters IC-Pak Anion HC Column, 4.6 mm ID, 150 mm Length, 10 μm Particle Size
Catalog No. 50785254 Shop All Waters Corporation Products
Encompass_Preferred
This column provides accurate and reproducible anion and cation analyses across a wide range of sample matrices and concentrations.
  • Enables versatile analysis of ions from simple to complex matrices.
  • Offers an exceptional linear loading range (less than 1.0 ppb to over 400 ppm).
  • No pH limitation on eluent or sample.
  • Separates and quantifies F, Cl, NO2, Br, NO3, HPO42, and SO42.
  • Recommended for inorganic ion analysis.
  • Compatible with standard laboratory chromatography equipment.
  • Supports high sensitivity conductivity detection and high throughput.
Catalog No. 50-785-254 Supplier Waters Corp Supplier No. WAT026770
This column provides accurate and reproducible anion and cation analyses across a wide range of sample matrices and concentrations.
  • Enables versatile analysis of ions from simple to complex matrices.
  • Offers an exceptional linear loading range (less than 1.0 ppb to over 400 ppm).
  • No pH limitation on eluent or sample.
  • Separates and quantifies F, Cl, NO2, Br, NO3, HPO42, and SO42.
  • Recommended for inorganic ion analysis.
  • Compatible with standard laboratory chromatography equipment.
  • Supports high sensitivity conductivity detection and high throughput.