Learn More
Waters Corp Waters IC-Pak Cation Guard Column, 10 μm, 4.6 mm × 50 mm, HPLC
Catalog No. 50784963 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters offers a range of products for ion chromatography. The IC-Pak resin based columns, including this one, are designed for analyzing various ions from diverse sample matrices, both simple and complex.
  • Allow the analysis of a full range of ions from numerous sample matrices, both simple and complex.
  • Offer an exceptional linear loading range of less than 1.0 ppb to greater than 400 ppm without dilution and without pH limitation on eluent or sample.
  • Flexibility exists for accurate and reproducible anion and cation analyses at all concentration levels.
Catalog No. 50-784-963 Supplier Waters Corp Supplier No. WAT007356
Waters offers a range of products for ion chromatography. The IC-Pak resin based columns, including this one, are designed for analyzing various ions from diverse sample matrices, both simple and complex.
  • Allow the analysis of a full range of ions from numerous sample matrices, both simple and complex.
  • Offer an exceptional linear loading range of less than 1.0 ppb to greater than 400 ppm without dilution and without pH limitation on eluent or sample.
  • Flexibility exists for accurate and reproducible anion and cation analyses at all concentration levels.