Learn More
Metrohm USA Zirconium oxide pump piston, high-performance, compatible with standard Metrohm pump heads, 6.2824.070
SDP
Catalog No. NC2234077 Shop All Metrohm USA Products
Encompass
The Metrohm 62824070 Zirconium oxide piston is a high-performance component designed for reliable and precise fluid delivery in Metrohm's ion chromatography (IC) systems. This pump piston, made from robust zirconium oxide, ensures exceptional durability and chemical resistance for demanding analytical applications. It is compatible with all standard Metrohm pump heads, simplifying maintenance and replacement, thereby optimizing instrument uptime.
  • Engineered for ion chromatography applications.
  • Made from durable zirconium oxide.
  • Facilitates precise fluid delivery in high-pressure pump heads.
  • Compatible with all standard Metrohm pump heads for easy maintenance.
  • Resistant to wear and chemicals.
  • Stable baseline for accurate measurements in various applications.
  • Serves as a replacement part for enhanced system reliability.
This item is not returnable. View return policy
Catalog No. NC2234077 Supplier Metrohm USA Supplier No. 62824070
The Metrohm 62824070 Zirconium oxide piston is a high-performance component designed for reliable and precise fluid delivery in Metrohm's ion chromatography (IC) systems. This pump piston, made from robust zirconium oxide, ensures exceptional durability and chemical resistance for demanding analytical applications. It is compatible with all standard Metrohm pump heads, simplifying maintenance and replacement, thereby optimizing instrument uptime.
  • Engineered for ion chromatography applications.
  • Made from durable zirconium oxide.
  • Facilitates precise fluid delivery in high-pressure pump heads.
  • Compatible with all standard Metrohm pump heads for easy maintenance.
  • Resistant to wear and chemicals.
  • Stable baseline for accurate measurements in various applications.
  • Serves as a replacement part for enhanced system reliability.