Learn More
CritiCore Protective Wear IC03 Non-Linting Cleanroom Coverall
Catalog No. 17001088
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-088 2X-Large
17-001-089 3X-Large
17-001-090 4X-Large
17-001-091 5X-Large
17-001-083 X-Small
17-001-084 Small
17-001-085 Medium
17-001-086 Large
17-001-087 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 17-001-088 Supplier CritiCore Protective Wear Supplier No. 13C26080IC03012XLWHT
Only null left
Add to Cart
Add to Cart
This IC03 non-linting cleanroom frock is designed to restrict particulates in controlled environments, and provides excellent ESD properties. This fabric has been engineered with a filament polyester rated for ISO class 5 cleanrooms and above.
  • IC03 Fabric
  • Grid-ESD / Non-Linting
  • Knit-Cuff Sleeves
  • Coil Zipper Front Enclosure
  • Neck Snaps at top of Mandarin Collar
  • Primary Application: Microelectronics, Semi-Conductor, Precision Optics, Wafer Manufacturing
Specifications
Product Type Cleanroom Coveralls
Size 2X-Large
Gender Unisex
Material Polyester
Color White
Disposable Reusable
Ankle Style Elastic
Cuff Style Knit
Neck Style Collared
Closure Type Zipper
Seam Type Stitched
Sterility Non-sterile
Resistance Particle