Learn More
CritiCore Protective Wear IC03 Non-Linting Cleanroom Coverall
Catalog No. 17604268
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-604-268 6X-Large
17-604-259 X-Small
17-604-260 Small
17-604-261 Medium
17-604-262 Large
17-604-263 X-Large
17-604-264 2X-Large
17-604-265 3X-Large
17-604-266 4X-Large
17-604-267 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 17-604-268 Supplier CritiCore Protective Wear Supplier No. 13C23065IC0301WHT6XL
Add to Cart
Add to Cart
This item is not returnable. View return policy
IC03 Non-Linting Cleanroom Coverall designed to restrict particulates in controlled environments, and provides excellent ESD properties. This fabric has been engineered with a filament polyester rated for ISO Class 5 Cleanrooms and Above.
  • IC03 Fabric
  • Grid-ESD / Non-Linting
  • Knit-Cuff Sleeves
  • Coil Zipper Front Enclosure
  • Neck Snaps at top of Mandarin Collar
  • Primary Application: Microelectronics, Semi-Conductor, Precision Optics, Wafer Manufacturing
Specifications
Product Type Cleanroom Coveralls
Size 6X-Large
Gender Unisex
Material Polyester
Color White
Disposable Reusable
Ankle Style Elastic
Cuff Style Knit
Neck Style Collared
Closure Type Front Zipper with Snap
Seam Type Stitched
Sterility Non-sterile
Resistance Particle