Learn More
TARGETMOL CHEMICALS INC IC261 25MG
SDP
Catalog No. 502580812
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IC261 (SU-5607) is a novel inhibitor of CK1 triggers the mitotic checkpoint control. The IC50 of IC261 for CK1 was 16 uM and for Cdk5 is 4.5 mM. purity: 99%
Catalog No. 50-258-0812 Supplier TARGETMOL CHEMICALS INC Supplier No. T244025MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IC261 (SU-5607) is a novel inhibitor of CK1 triggers the mitotic checkpoint control. The IC50 of IC261 for CK1 was 16 uM and for Cdk5 is 4.5 mM. purity: 99%