Learn More
Julabo USA Inc IC40 Immersion Cooler -40C
SDP
Catalog No. 5000077872
Encompass_Preferred
Immersion Cooler JULABO IC40 115V/60Hz R290 Refrigerant -40 100 C Cooling 0 25kW at 20C 0 23kW at 0C 0 12kW at -20C 0 02kW at -40C Cooling cylinder 3 9cm x 2cm diameter on 47 2 cm tether 9 1x 17 3 x 15 7 in 46 3lbs The specification of the working temperature range above the ambient temperature corresponds to the maximum permissible temperature in the application for which the immersion cooler is approved The lowest temperature of the working temperature range corresponds to the minimum temperature that the probe reaches at its surface
Catalog No. 50-000-77872 Supplier Julabo USA Inc Supplier No. 9651040.3
Add to Cart
Add to Cart
Request a Quote
Immersion Cooler JULABO IC40 115V/60Hz R290 Refrigerant -40 100 C Cooling 0 25kW at 20C 0 23kW at 0C 0 12kW at -20C 0 02kW at -40C Cooling cylinder 3 9cm x 2cm diameter on 47 2 cm tether 9 1x 17 3 x 15 7 in 46 3lbs The specification of the working temperature range above the ambient temperature corresponds to the maximum permissible temperature in the application for which the immersion cooler is approved The lowest temperature of the working temperature range corresponds to the minimum temperature that the probe reaches at its surface