Learn More
Julabo USA Inc IC50 Immersion Cooler -50C
SDP
Catalog No. 5000077885
Encompass_Preferred
Immersion Cooler JULABO IC50 115V/60Hz R290 Refrigerant -50 100 C Cooling 0 45kW at 20C 0 32kW at 0C 0 16kW at -20C 0 04kW at -40C Cooling cylinder 3 9cm x 2cm diameter on 47 2 cm tether 9 1x 17 3 x 15 7 in 47 4lbs The specification of the working temperature range above the ambient temperature corresponds to the maximum permissible temperature in the application for which the immersion cooler is approved The lowest temperature of the working temperature range corresponds to the minimum temperature that the probe reaches at its surface
Catalog No. 50-000-77885 Supplier Julabo USA Inc Supplier No. 9651050.30003826
Add to Cart
Add to Cart
Request a Quote
Immersion Cooler JULABO IC50 115V/60Hz R290 Refrigerant -50 100 C Cooling 0 45kW at 20C 0 32kW at 0C 0 16kW at -20C 0 04kW at -40C Cooling cylinder 3 9cm x 2cm diameter on 47 2 cm tether 9 1x 17 3 x 15 7 in 47 4lbs The specification of the working temperature range above the ambient temperature corresponds to the maximum permissible temperature in the application for which the immersion cooler is approved The lowest temperature of the working temperature range corresponds to the minimum temperature that the probe reaches at its surface