Learn More
Medchemexpress LLC HY-100457 10mg Medchemexpress, IC87201 CAS:866927-10-8 Purity:>98%
SDP
Catalog No. 502080589
Encompass_Preferred
Medchemexpress, HY-100457 10mg IC87201 CAS:866927-10-8 IC87201, an inhibitor of PSD95-nNOS protein-protein interactions, suppresses NMDAR-dependent NO and cGMP formation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0589 Supplier Medchemexpress LLC Supplier No. HY10045710MG
Medchemexpress, HY-100457 10mg IC87201 CAS:866927-10-8 IC87201, an inhibitor of PSD95-nNOS protein-protein interactions, suppresses NMDAR-dependent NO and cGMP formation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.