Learn More
Medchemexpress LLC ICA-105665 (PF-04895162) | 2694728-63-5 | 99.9% | 355.34 | C19H15F2N3O2 | 10MG
SDP
Catalog No. 5000239553
Encompass_Preferred
ICA-105665 is a research small molecule that acts as a neuronal Kv7.2/7.3 and Kv7.3/7.5 potassium channel opener. It is provided as a powder for in vitro and in vivo studies, and is intended for research use only. Store the solid at low temperature for long-term stability and follow standard handling practices for research chemicals.
  • Potent neuronal Kv7 channel opener for electrophysiology and pharmacology studies.
  • Suitable for in vitro and in vivo research applications.
  • High research-grade purity reported at about 99.9%.
  • Molecular weight 355.34 g/mol; chemical formula C19H15F2N3O2.
  • Stable when stored at recommended low temperatures.
Catalog No. 50-002-39553 Supplier Medchemexpress LLC Supplier No. HY12546910MG
ICA-105665 is a research small molecule that acts as a neuronal Kv7.2/7.3 and Kv7.3/7.5 potassium channel opener. It is provided as a powder for in vitro and in vivo studies, and is intended for research use only. Store the solid at low temperature for long-term stability and follow standard handling practices for research chemicals.
  • Potent neuronal Kv7 channel opener for electrophysiology and pharmacology studies.
  • Suitable for in vitro and in vivo research applications.
  • High research-grade purity reported at about 99.9%.
  • Molecular weight 355.34 g/mol; chemical formula C19H15F2N3O2.
  • Stable when stored at recommended low temperatures.