Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ICA-105665 (PF-04895162) | 2694728-63-5 | 99.9% | 355.34 | C19H15F2N3O2 | 25MG
SDP

Catalog No. 5000235692
Encompass_Preferred

ICA-105665 is an orally active small-molecule opener of neuronal Kv7 potassium channels (Kv7.2/7.3 and Kv7.3/7.5) used in research. It penetrates the blood-brain barrier, exhibits antiseizure activity, and has reported effects on liver mitochondrial function and bile salt export protein (BSEP) transport (IC50 311 μM). The compound is supplied as a high-purity white to off-white powder suitable for in vitro and in vivo studies.

  • Cas registry number: 2694728-63-5.
  • Mechanism of action: opens Kv7.2/7.3 and Kv7.3/7.5 potassium channels.
  • Chemical formula: C19H15F2N3O2.
  • Molecular weight: 355.34 g/mol.
  • Purity: 99.9% (rounded).
  • Appearance and form: white to off-white powder.
  • Solubility: soluble in DMSO at 250 mg/mL; ultrasonic assistance recommended.
  • Storage: store powder at -20°C for long-term stability; store solutions at -80°C for short-term use.

Catalog No. 50-002-35692 Supplier Medchemexpress LLC Supplier No. HY12546925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ICA-105665 is an orally active small-molecule opener of neuronal Kv7 potassium channels (Kv7.2/7.3 and Kv7.3/7.5) used in research. It penetrates the blood-brain barrier, exhibits antiseizure activity, and has reported effects on liver mitochondrial function and bile salt export protein (BSEP) transport (IC50 311 μM). The compound is supplied as a high-purity white to off-white powder suitable for in vitro and in vivo studies.

  • Cas registry number: 2694728-63-5.
  • Mechanism of action: opens Kv7.2/7.3 and Kv7.3/7.5 potassium channels.
  • Chemical formula: C19H15F2N3O2.
  • Molecular weight: 355.34 g/mol.
  • Purity: 99.9% (rounded).
  • Appearance and form: white to off-white powder.
  • Solubility: soluble in DMSO at 250 mg/mL; ultrasonic assistance recommended.
  • Storage: store powder at -20°C for long-term stability; store solutions at -80°C for short-term use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.