Learn More
Medchemexpress LLC ICA-105665 (PF-04895162) | 2694728-63-5 | 99.9% | 355.34 | C19H15F2N3O2 | 5MG
SDP
Catalog No. 5000236318
Encompass_Preferred
ICA-105665 is a small-molecule research compound that acts as an orally active neuronal Kv7 potassium channel opener selective for Kv7.2/7.3 and Kv7.3/7.5. It is provided for laboratory research in milligram-scale quantities and is available as a solid or as a 10 mM solution in DMSO. Molecular formula C19H15F2N3O2; molecular weight 355.34.
Catalog No. 50-002-36318 Supplier Medchemexpress LLC Supplier No. HY1254695MG
ICA-105665 is a small-molecule research compound that acts as an orally active neuronal Kv7 potassium channel opener selective for Kv7.2/7.3 and Kv7.3/7.5. It is provided for laboratory research in milligram-scale quantities and is available as a solid or as a 10 mM solution in DMSO. Molecular formula C19H15F2N3O2; molecular weight 355.34.