Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ICA 110381 | 325457-99-6 | MFCD05669575 | 99.9% | 267.11 g/mol | C12H8Cl2N2O | 5 MG
SDP

Catalog No. 5000263246
Encompass_Preferred

ICA 110381 is a small-molecule activator of Kv7.2/Kv7.3 (KCNQ2/3) potassium channels with reported anticonvulsant activity and a documented EC50 of ≈0.38 μM. It is supplied as a high-purity research chemical for use in studies of neuronal excitability and ion channel pharmacology.

  • Potent Kv7.2/Kv7.3 activator (EC50 ≈ 0.38 μM).
  • High purity suitable for research applications.
  • Demonstrated anticonvulsant activity in vivo.
  • Soluble in DMSO; store protected from light at recommended temperatures.
  • Molecular weight 267.11 g/mol; formula C12H8Cl2N2O.

Catalog No. 50-002-63246 Supplier Medchemexpress LLC Supplier No. HY1085875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ICA 110381 is a small-molecule activator of Kv7.2/Kv7.3 (KCNQ2/3) potassium channels with reported anticonvulsant activity and a documented EC50 of ≈0.38 μM. It is supplied as a high-purity research chemical for use in studies of neuronal excitability and ion channel pharmacology.

  • Potent Kv7.2/Kv7.3 activator (EC50 ≈ 0.38 μM).
  • High purity suitable for research applications.
  • Demonstrated anticonvulsant activity in vivo.
  • Soluble in DMSO; store protected from light at recommended temperatures.
  • Molecular weight 267.11 g/mol; formula C12H8Cl2N2O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.