Learn More
TARGETMOL CHEMICALS INC ICA-121431 25MG
SDP
Catalog No. 502579213
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ICA-121431 (22-Diphenyl-N-[4-(thiazol-2-ylsulfamoyl) is a nanomolar potent small molecule Nav1.7 channel inhibitor with IC50 of 19 nM for rat Nav1.7 with little or no activity against human Nav1.5 or Nav1.7 channels. purity: 98%
Catalog No. 50-257-9213 Supplier TARGETMOL CHEMICALS INC Supplier No. T733625MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ICA-121431 (22-Diphenyl-N-[4-(thiazol-2-ylsulfamoyl) is a nanomolar potent small molecule Nav1.7 channel inhibitor with IC50 of 19 nM for rat Nav1.7 with little or no activity against human Nav1.5 or Nav1.7 channels. purity: 98%