Learn More
Medchemexpress LLC ICA Mouse HEK293 10ug
SDP
Catalog No. 5000179325
Encompass_Preferred
ICA Protein a potent inhibitor of carbonic anhydrase 2 (CA2) intricately regulates enzymatic activity within a finely tuned network Functionally ICA a non-iron-binding monomer exhibits specificity in inhibiting CA2 Its transferrin-like domain 2 engages in molecular interactions underscoring ICA s regulatory role This positions ICA as a key player adding nuance to the precise mechanisms governing carbonic anhydrase activity ICA Protein Mouse (HEK293 His) is the recombinant mouse-derived ICA protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-79325 Supplier Medchemexpress LLC Supplier No. HYP7391210UG
ICA Protein a potent inhibitor of carbonic anhydrase 2 (CA2) intricately regulates enzymatic activity within a finely tuned network Functionally ICA a non-iron-binding monomer exhibits specificity in inhibiting CA2 Its transferrin-like domain 2 engages in molecular interactions underscoring ICA s regulatory role This positions ICA as a key player adding nuance to the precise mechanisms governing carbonic anhydrase activity ICA Protein Mouse (HEK293 His) is the recombinant mouse-derived ICA protein expressed by HEK293 with C-His labeled tag