Learn More
Aviva Systems Biology Corp ICA1L Antibody - middle region (ARP52622_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52622_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ICA1L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ICA1L. Alternate names: ALS2CR14, ALS2CR15, DKFZp434E1919, MGC138440ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60787